siRNA Details
Gene Symbol: Map3k14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108301.1 | NP_001101771.1 | 9 | 1712 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Map3k14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108301.1 | NP_001101771.1 | 9 | 1712 |