siRNA Details
| Gene Symbol: Map3k14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108301.1 | NP_001101771.1 | 13 | 2442 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map3k14| Gene Symbol: Map3k14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108301.1 | NP_001101771.1 | 13 | 2442 | ||