siRNA Details
| Gene Symbol: Kansl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768297.1 | XP_008766519.1 | 2 | 1359 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kansl1| Gene Symbol: Kansl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768297.1 | XP_008766519.1 | 2 | 1359 | ||