siRNA Details
| Gene Symbol: RGD1359290 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047898.1 | NP_001041363.1 | 5, 6 | 556 | ||
| XM_008768380.1 | XP_008766602.1 | 3, 4 | 661 | |||
| GenBank mRNA | AY321338.1 | - | 5, 6 | 556 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1359290| Gene Symbol: RGD1359290 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047898.1 | NP_001041363.1 | 5, 6 | 556 | ||
| XM_008768380.1 | XP_008766602.1 | 3, 4 | 661 | |||
| GenBank mRNA | AY321338.1 | - | 5, 6 | 556 | ||