siRNA Details
Gene Symbol: Sdk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108303.2 | NP_001101773.1 | 38 | 5359 | ||
XM_006247693.2 | XP_006247755.1 | 41 | 6886 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sdk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108303.2 | NP_001101773.1 | 38 | 5359 | ||
XM_006247693.2 | XP_006247755.1 | 41 | 6886 |