siRNA Details
| Gene Symbol: Sdk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108303.2 | NP_001101773.1 | 19 | 2599 | ||
| XM_006247693.2 | XP_006247755.1 | 21 | 4069 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sdk2| Gene Symbol: Sdk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108303.2 | NP_001101773.1 | 19 | 2599 | ||
| XM_006247693.2 | XP_006247755.1 | 21 | 4069 | |||