siRNA Details
Gene Symbol: Gga3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108304.1 | NP_001101774.1 | 16 | 2936 | ||
XM_006247719.2 | XP_006247781.1 | 17 | 2122 | |||
XM_006247720.2 | XP_006247782.1 | 15 | 2260 | |||
XM_006247721.1 | XP_006247783.1 | 16 | 1946 |
