siRNA Details
| Gene Symbol: Ten1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108307.1 | NP_001101777.1 | 4 | 577 | ||
| GenBank mRNA | BC168942.1 | - | 3 | 406 | 8365819 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ten1| Gene Symbol: Ten1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108307.1 | NP_001101777.1 | 4 | 577 | ||
| GenBank mRNA | BC168942.1 | - | 3 | 406 | 8365819 | |