siRNA Details
| Gene Symbol: Jmjd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012143.2 | NP_001012143.2 | 2 | 688 | ||
| XM_006247806.1 | XP_006247868.1 | 2 | 521 | |||
| GenBank mRNA | BC079012.1 | - | 2 | 688 | 7113380 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jmjd6| Gene Symbol: Jmjd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012143.2 | NP_001012143.2 | 2 | 688 | ||
| XM_006247806.1 | XP_006247868.1 | 2 | 521 | |||
| GenBank mRNA | BC079012.1 | - | 2 | 688 | 7113380 | |