siRNA Details
| Gene Symbol: Mfsd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108308.1 | NP_001101778.1 | 4 | 576 | ||
| XM_006247809.2 | XP_006247871.1 | 5 | 924 | |||
| XM_006247810.2 | XP_006247872.1 | 5 | 525 | |||
| XM_006247811.2 | XP_006247873.1 | 3 | 257 | |||

Mfsd11
Genomic Map