siRNA Details
| Gene Symbol: Enthd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100729.1 | NP_001094199.1 | 13 | 1715 | ||
| XM_006247893.2 | XP_006247955.1 | 14 | 1741 | |||
| XM_006247894.2 | XP_006247956.1 | 13 | 1850 | |||
| XM_006247895.2 | XP_006247957.1 | 13 | 1741 | |||
| XM_008768478.1 | XP_008766700.1 | 13 | 1924 | |||
| GenBank mRNA | BC089956.1 | - | 4 | 654 | ||

Enthd2
Genomic Map