siRNA Details
Gene Symbol: Narf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039207.2 | NP_001034296.1 | 5 | 506 | ||
XM_006247899.2 | XP_006247961.1 | 5 | 539 | |||
GenBank mRNA | BC110050.1 | - | 5 | 506 | 7454395 |
