siRNA Details
| Gene Symbol: Wdr45b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039587.1 | NP_001034676.1 | 7, 8 | 712 | ||
| XM_006247900.2 | XP_006247962.1 | 7, 8 | 855 | |||
| GenBank mRNA | AM182327.1 | - | 7, 8 | 712 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr45b| Gene Symbol: Wdr45b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039587.1 | NP_001034676.1 | 7, 8 | 712 | ||
| XM_006247900.2 | XP_006247962.1 | 7, 8 | 855 | |||
| GenBank mRNA | AM182327.1 | - | 7, 8 | 712 | ||