siRNA Details
| Gene Symbol: Tbcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271364.1 | NP_001258293.1 | 5 | 577 | ||
| XM_006247861.2 | XP_006247923.1 | 5 | 638 | |||
| XM_006247864.2 | XP_006247926.1 | 5 | 638 | |||
| XM_008768500.1 | XP_008766722.1 | 5 | 638 | |||
| XM_008768501.1 | XP_008766723.1 | 5 | 581 | |||

Tbcd
Genomic Map