siRNA Details
| Gene Symbol: LOC498829 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY325193.1 | - | 5 | 922 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC498829| Gene Symbol: LOC498829 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY325193.1 | - | 5 | 922 | ||