siRNA Details
| Gene Symbol: Dscr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108316.2 | NP_001101786.2 | 3 | 358 | ||
| XM_006248073.2 | XP_006248135.1 | 3 | 811 | |||
| XM_008768583.1 | XP_008766805.1 | 3 | 812 | |||
| GenBank mRNA | BC160826.1 | - | 3 | 358 | 7444837 | |

Dscr3
Genomic Map