siRNA Details
| Gene Symbol: Hmgn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013184.1 | NP_001013202.1 | 5 | 233 | ||
| GenBank mRNA | BC086423.1 | - | 5 | 233 | 7302011 | |
| FQ220876.1 | - | 5 | 271 | |||
| FQ222121.1 | - | 5 | 272 | |||
| FQ230156.1 | - | 5 | 236 | |||
| Gene Symbol: LOC100911295 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751029.3 | XP_003751077.1 | 5 | 304 | ||

Hmgn1
Genomic Map