siRNA Details
Gene Symbol: Tfg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012144.1 | NP_001012144.1 | 7 | 1059 | ||
XM_006248236.2 | XP_006248298.1 | 7 | 1173 | |||
XM_006248237.2 | XP_006248299.1 | 7 | 1128 | |||
GenBank mRNA | AB218900.1 | - | 6 | 800 | ||
BC078947.1 | - | 7 | 1059 | 7109719 |
