siRNA Details
| Gene Symbol: Gtpbp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025015.1 | NP_001020186.1 | 4, 5 | 681 | ||
| XM_006248340.2 | XP_006248402.1 | 3, 4 | 607 | |||
| GenBank mRNA | BC091236.1 | - | 4, 5 | 681 | 7322967 | |
| FQ216550.1 | - | 4, 5 | 680 | |||

Gtpbp8
Genomic Map