siRNA Details
| Gene Symbol: Fam162a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029903.1 | NP_001025074.1 | 2, 3 | 156 | ||
| XM_006248413.2 | XP_006248475.1 | 2, 3 | 244 | |||
| GenBank mRNA | BC097971.1 | - | 2, 3 | 190 | 7442058 | |
| FQ222091.1 | - | 2, 3 | 214 | |||
| M17412.1 | - | 1, 2 | 129 | |||

Fam162a
Genomic Map