siRNA Details
| Gene Symbol: Fam162a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029903.1 | NP_001025074.1 | 4 | 351 | ||
| XM_006248413.2 | XP_006248475.1 | 4 | 439 | |||
| GenBank mRNA | BC097971.1 | - | 4 | 385 | 7442058 | |
| FQ222091.1 | - | 4 | 409 | |||
| M17412.1 | - | 3 | 324 | |||

Fam162a
Genomic Map