siRNA Details
| Gene Symbol: Pdia5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014125.1 | NP_001014147.1 | 4 | 344 | ||
| GenBank mRNA | BC088305.1 | - | 4 | 344 | 7323988 | |
| Gene Symbol: LOC100911928 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248418.2 | XP_006248480.1 | 4 | 435 | ||

Pdia5
Genomic Map