siRNA Details
| Gene Symbol: SNX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003794.3 | NP_003785.1 | 4 | 509 | ||
| NR_073435.1 | 3 | 387 | ||||
| XM_017007414.1 | XP_016862903.1 | 4 | 501 | |||
| GenBank mRNA | AF065485.1 | - | 4 | 467 | ||
| AK001835.1 | - | 4 | 471 | |||
| AK298972.1 | - | 3 | 354 | |||
| BC018762.1 | - | 4 | 485 | 4641014 | ||
| BI669801.1 | - | 4 | 513 | |||
| DA289870.1 | - | 4 | 511 | |||

SNX4
Genomic Map