siRNA Details
| Gene Symbol: Rtp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108321.1 | NP_001101791.1 | 4 | 935 | ||
| XM_006248521.2 | XP_006248583.1 | 2 | 1322 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rtp4| Gene Symbol: Rtp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108321.1 | NP_001101791.1 | 4 | 935 | ||
| XM_006248521.2 | XP_006248583.1 | 2 | 1322 | |||