siRNA Details
| Gene Symbol: Tbx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108322.1 | NP_001101792.1 | 5, 6 | 790 | ||
| XM_006248619.2 | XP_006248681.1 | 4, 5 | 1572 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbx1| Gene Symbol: Tbx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108322.1 | NP_001101792.1 | 5, 6 | 790 | ||
| XM_006248619.2 | XP_006248681.1 | 4, 5 | 1572 | |||