siRNA Details
| Gene Symbol: Dgcr14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012472.1 | NP_001012490.1 | 6 | 720 | ||
| XM_006248635.2 | XP_006248697.1 | 6 | 765 | |||
| GenBank mRNA | BC089898.1 | - | 6 | 720 | 7382217 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dgcr14| Gene Symbol: Dgcr14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012472.1 | NP_001012490.1 | 6 | 720 | ||
| XM_006248635.2 | XP_006248697.1 | 6 | 765 | |||
| GenBank mRNA | BC089898.1 | - | 6 | 720 | 7382217 | |