siRNA Details
| Gene Symbol: Dgcr14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012472.1 | NP_001012490.1 | 1, 2 | 152 | ||
| XM_006248635.2 | XP_006248697.1 | 1, 2 | 200 | |||
| XM_006248636.2 | XP_006248698.1 | 1, 2 | 200 | |||
| GenBank mRNA | BC089898.1 | - | 1, 2 | 152 | 7382217 | |

Dgcr14
Genomic Map