siRNA Details
| Gene Symbol: Lztr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108326.2 | NP_001101796.1 | 15, 16 | 1823 | ||
| XM_006248707.2 | XP_006248769.1 | 15, 16 | 2136 | |||
| GenBank mRNA | BC169071.1 | - | 15, 16 | 1823 | 7938530 | |
| FQ211947.1 | - | 15, 16 | 1878 | |||

Lztr1
Genomic Map