siRNA Details
| Gene Symbol: Ppil2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017383.1 | NP_001017383.1 | 9 | 535 | ||
| XM_008768892.1 | XP_008767114.1 | 9 | 571 | |||
| GenBank mRNA | BC085890.1 | - | 9 | 535 | 7192457 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppil2| Gene Symbol: Ppil2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017383.1 | NP_001017383.1 | 9 | 535 | ||
| XM_008768892.1 | XP_008767114.1 | 9 | 571 | |||
| GenBank mRNA | BC085890.1 | - | 9 | 535 | 7192457 | |