siRNA Details
Gene Symbol: Spag6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034960.1 | NP_001030132.1 | 3, 4 | 337 | ||
GenBank mRNA | BC104678.1 | - | 3, 4 | 337 | 7454069 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Spag6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034960.1 | NP_001030132.1 | 3, 4 | 337 | ||
GenBank mRNA | BC104678.1 | - | 3, 4 | 337 | 7454069 |