siRNA Details
| Gene Symbol: Prkdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751100.3 | XP_003751148.1 | 2, 3 | 498 | ||
| XM_008768933.1 | XP_008767155.1 | 2, 3 | 499 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkdc| Gene Symbol: Prkdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751100.3 | XP_003751148.1 | 2, 3 | 498 | ||
| XM_008768933.1 | XP_008767155.1 | 2, 3 | 499 | |||