siRNA Details
| Gene Symbol: Zfp358 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108328.1 | NP_001101798.1 | 2 | 1756 | ||
| XM_006248767.1 | XP_006248829.1 | 2 | 1663 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp358| Gene Symbol: Zfp358 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108328.1 | NP_001101798.1 | 2 | 1756 | ||
| XM_006248767.1 | XP_006248829.1 | 2 | 1663 | |||