siRNA Details
| Gene Symbol: Medag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751115.3 | XP_003751163.1 | 1100 | |||
| XM_341029.6 | XP_341030.3 | 1376 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Medag| Gene Symbol: Medag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751115.3 | XP_003751163.1 | 1100 | |||
| XM_341029.6 | XP_341030.3 | 1376 | ||||