siRNA Details
| Gene Symbol: Zfp157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170404.1 | NP_001163875.1 | 5 | 1131 | ||
| XM_008769063.1 | XP_008767285.1 | 7 | 1348 | |||
| XM_008769064.1 | XP_008767286.1 | 7 | 1658 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp157| Gene Symbol: Zfp157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170404.1 | NP_001163875.1 | 5 | 1131 | ||
| XM_008769063.1 | XP_008767285.1 | 7 | 1348 | |||
| XM_008769064.1 | XP_008767286.1 | 7 | 1658 | |||