siRNA Details
| Gene Symbol: Wbscr16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108332.1 | NP_001101802.1 | 8, 9 | 1105 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wbscr16| Gene Symbol: Wbscr16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108332.1 | NP_001101802.1 | 8, 9 | 1105 | ||