siRNA Details
| Gene Symbol: Snrnp35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014127.1 | NP_001014149.1 | 2 | 577 | ||
| XM_006249325.2 | XP_006249387.1 | 2 | 625 | |||
| GenBank mRNA | BC086435.1 | - | 2 | 577 | 7302221 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snrnp35| Gene Symbol: Snrnp35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014127.1 | NP_001014149.1 | 2 | 577 | ||
| XM_006249325.2 | XP_006249387.1 | 2 | 625 | |||
| GenBank mRNA | BC086435.1 | - | 2 | 577 | 7302221 | |