siRNA Details
| Gene Symbol: Rsrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014128.1 | NP_001014150.1 | 8 | 904 | ||
| XM_006249338.2 | XP_006249400.1 | 6 | 771 | |||
| XM_006249339.2 | XP_006249401.1 | 6 | 771 | |||
| XM_006249340.2 | XP_006249402.1 | 7 | 862 | |||
| XM_006249341.2 | XP_006249403.1 | 7 | 862 | |||
| GenBank mRNA | BC087067.1 | - | 8 | 904 | 7133034 | |

Rsrc2
Genomic Map