siRNA Details
| Gene Symbol: Rasal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108335.1 | NP_001101805.1 | 4 | 416 | ||
| XM_006249395.2 | XP_006249457.1 | 3 | 1613 | |||
| XM_006249396.2 | XP_006249458.1 | 3 | 1622 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rasal1| Gene Symbol: Rasal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108335.1 | NP_001101805.1 | 4 | 416 | ||
| XM_006249395.2 | XP_006249457.1 | 3 | 1613 | |||
| XM_006249396.2 | XP_006249458.1 | 3 | 1622 | |||