siRNA Details
| Gene Symbol: Ddx54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191548.1 | NP_001178477.1 | 5 | 646 | ||
| XM_006249397.1 | XP_006249459.1 | 5 | 722 | |||
| XM_006249398.1 | XP_006249460.1 | 5 | 722 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddx54| Gene Symbol: Ddx54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191548.1 | NP_001178477.1 | 5 | 646 | ||
| XM_006249397.1 | XP_006249459.1 | 5 | 722 | |||
| XM_006249398.1 | XP_006249460.1 | 5 | 722 | |||