siRNA Details
| Gene Symbol: Sdsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108336.1 | NP_001101806.1 | 7 | 979 | ||
| XM_006249399.2 | XP_006249461.1 | 5 | 960 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sdsl| Gene Symbol: Sdsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108336.1 | NP_001101806.1 | 7 | 979 | ||
| XM_006249399.2 | XP_006249461.1 | 5 | 960 | |||