siRNA Details
| Gene Symbol: Med13l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006249420.2 | XP_006249482.1 | 26 | 5798 | ||
| XM_006249421.2 | XP_006249483.1 | 27 | 6175 | |||
| XM_006249422.2 | XP_006249484.1 | 27 | 6148 | |||
| GenBank mRNA | GU566026.1 | - | 27 | 6136 | ||

Med13l
Genomic Map