siRNA Details
| Gene Symbol: Fbxo21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CB615901.1 | - | 1 | 120 | ||
| CB715798.1 | - | 3 | 443 | |||
| FQ212189.1 | - | 10 | 1518 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo21| Gene Symbol: Fbxo21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CB615901.1 | - | 1 | 120 | ||
| CB715798.1 | - | 3 | 443 | |||
| FQ212189.1 | - | 10 | 1518 | |||