siRNA Details
| Gene Symbol: Pxn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012147.1 | NP_001012147.1 | 7 | 1339 | ||
| XM_006249455.2 | XP_006249517.1 | 12 | 2761 | |||
| XM_006249457.2 | XP_006249519.1 | 9 | 2104 | |||
| XM_006249459.2 | XP_006249521.1 | 8 | 2001 | |||
| XM_006249460.2 | XP_006249522.1 | 8 | 1258 | |||
| GenBank mRNA | AY641810.1 | - | 8 | 1253 | ||
| BC081984.1 | - | 7 | 1339 | 7128801 | ||
| FQ226157.1 | - | 8 | 1254 | |||

Pxn
Genomic Map