siRNA Details
| Gene Symbol: LOC100909836 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751199.3 | XP_003751247.1 | 2 | 436 | ||
| Gene Symbol: Gatc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108339.1 | NP_001101809.1 | 398 | |||
| XM_008769339.1 | XP_008767561.1 | 436 | ||||

LOC100909836
Genomic Map