siRNA Details
| Gene Symbol: Sppl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751200.3 | XP_003751248.1 | 4, 5 | 416 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sppl3| Gene Symbol: Sppl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751200.3 | XP_003751248.1 | 4, 5 | 416 | ||