siRNA Details
| Gene Symbol: Hscb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108340.1 | NP_001101810.1 | 4 | 488 | ||
| XM_006249564.2 | XP_006249626.1 | 4 | 520 | |||
| XM_008769340.1 | XP_008767562.1 | 4 | 521 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hscb| Gene Symbol: Hscb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108340.1 | NP_001101810.1 | 4 | 488 | ||
| XM_006249564.2 | XP_006249626.1 | 4 | 520 | |||
| XM_008769340.1 | XP_008767562.1 | 4 | 521 | |||