siRNA Details
| Gene Symbol: Noc4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014129.1 | NP_001014151.1 | 10, 11 | 1001 | ||
| GenBank mRNA | BC088275.1 | - | 10, 11 | 1001 | 7380687 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Noc4l| Gene Symbol: Noc4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014129.1 | NP_001014151.1 | 10, 11 | 1001 | ||
| GenBank mRNA | BC088275.1 | - | 10, 11 | 1001 | 7380687 | |