siRNA Details
| Gene Symbol: Wbscr22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135743.1 | NP_001129215.1 | 8, 9 | 619 | ||
| GenBank mRNA | BC160891.1 | - | 8, 9 | 619 | 7442902 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wbscr22| Gene Symbol: Wbscr22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135743.1 | NP_001129215.1 | 8, 9 | 619 | ||
| GenBank mRNA | BC160891.1 | - | 8, 9 | 619 | 7442902 | |