siRNA Details
| Gene Symbol: Abhd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271180.1 | NP_001258109.1 | 5 | 768 | ||
| GenBank mRNA | BC088239.1 | - | 4 | 893 | ||
| FQ213935.1 | - | 5 | 759 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abhd11| Gene Symbol: Abhd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271180.1 | NP_001258109.1 | 5 | 768 | ||
| GenBank mRNA | BC088239.1 | - | 4 | 893 | ||
| FQ213935.1 | - | 5 | 759 | |||