siRNA Details
| Gene Symbol: Lypd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007727.1 | NP_001007728.1 | 3 | 1157 | ||
| GenBank mRNA | BC082032.1 | - | 3 | 1157 | 7130639 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lypd1| Gene Symbol: Lypd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007727.1 | NP_001007728.1 | 3 | 1157 | ||
| GenBank mRNA | BC082032.1 | - | 3 | 1157 | 7130639 | |